Ammonium nitrate ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Warning

31114-500G 31114-2.5KG 31114-6X1KG 31114-1KG
BJ31114-2.5KGEA 187.41 USD
BJ31114-2.5KG BJ31114-1KG BJ31114-500G BJ31114-6X1KG
Ammonium nitrate ≥98%, Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Ammonium nitrate
Formula: NH₄NO₃
MW: 80.04 g/mol
Melting Pt: 169 °C.(1.013 hPa).170 °C
Density: 1.72 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011425
CAS Number: 6484-52-2
EINECS: 229-347-8
UN: 1942
ADR: 5.1,III

Order Now

Specification Test Results

Assay Min. 98.0 %
Water insoluble matter Max. 0.005 %
Residue on ignition (SO₄) Max. 0.01 %
pH (5 %, 25 °C) 4.5 - 6.0
Calcium (Ca) Max. 0.002 %
Cadmium (Cd) Max. 0.0005 %
Copper (Cu) Max. 0.0005 %
Iron (Fe) Max. 1 ppm
Potassium (K) Max. 0.001 %
Magnesium (Mg) Max. 0.001 %
Sodium (Na) Max. 0.001 %
Lead (Pb) Max. 0.0005 %
Zinc (Zn) Max. 0.0005 %
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 3 ppm
Nitrite (NO₂) Max. 0.0005 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.002 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR