Ammonium iron (II) sulfate hexahydrate ≥99.0% (RT), Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Ferrous Ammonium sulphate Hexahydrate
, Ferrous ammonium sulfate hexahydrate
, Ammonium iron(Ⅱ) sulfate hexahydrate
, Ammonium ferrous sulphate hexahydrate
, Ferrous Ammonium sulfate Hexahydrate
, di-Ammonium iron (II) sulphate hexahydrate
, Ammonium ferrous sulfate hexahydrate
, Ferrous ammonium sulphate hexahydrate
, Ammonium iron(II) sulphate hydrate
, di-Ammonium iron (II) sulfate hexahydrate
, Ammonium iron(Ⅱ) sulphate hexahydrate
, Ammonium iron(II) sulfate hydrate
09720-1KG
BJ09720-1KGEA
165.09
USD
BJ09720-1KG
Ammonium iron (II) sulfate hexahydrate ≥99.0% (RT), Puriss. p.a. ISO, Reag. Ph. Eur., Fluka™
Ammonium iron (II) sulphate hexahydrate
Formula:
(NH₄)₂Fe(SO₄)₂·6H₂O MW: 392.14 g/mol Melting Pt: 100 °C (dec.) Density: 1.86 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00150530 CAS Number: 7783-85-9 EINECS: 233-151-8 |
Specification Test Results
Assay (manganometric) | Min. 99.0 % |
pH (5 %, 25 °C) | 3.0 - 5.0 |
Calcium (Ca) | Max. 20 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 20 ppm |
Chromium (Cr) | Max. 50 ppm |
Copper (Cu) | Max. 20 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 100 ppm |
Manganese (Mn) | Max. 500 ppm |
Sodium (Na) | Max. 50 ppm |
Nickel (Ni) | Max. 200 ppm |
Lead (Pb) | Max. 10 ppm |
Zinc (Zn) | Max. 20 ppm |
Iron(III) | Max. 200 ppm |
Chloride (Cl) | Max. 10 ppm |
Phosphate (PO₄) | Max. 20 ppm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...