Ammonium carbonate ≥30,0% (NH₃ basis) ACS, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Salt of hartshorn

207861-100G 207861-500G
BJ207861-100GEA 83 USD
BJ207861-100G BJ207861-2.5KG BJ207861-25G BJ207861-500G
Ammonium carbonate ≥30,0% (NH₃ basis) ACS, Fluka™
Ammonium carbonate
Formula: (NH₄)₂CO₃
Melting Pt: >59 °C
Density: 0.9 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010890
CAS Number: 506-87-6
EINECS: 208-058-0

Order Now

Specification Test Results

Assay of NH₃ Min. 30.0 %
Non-volatile matter Max. 0.01 %
Water insoluble matter Max. 0.005 %
Iron (Fe) Max. 5 ppm
Heavy metals (as Pb) Max. 5 ppm
Chloride (Cl) Max. 5 ppm
Total S (as SO₄) Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR