Ammonium dihydrogen phosphate ≥98% ACS, Fluka™
Supplier: Honeywell Research Chemicals
Synonyms:
Ammonium dihydrogenphosphate
, Ammonium phosphate, monobasic
, Ammonium Phosphate Monobasic
216003-100G
216003-2.5KG
216003-500G
BJ216003-2.5KGEA
403.44
USD
BJ216003-2.5KG
BJ216003-500G
BJ216003-100G
Ammonium dihydrogen phosphate ≥98% ACS, Fluka™
Ammonium dihydrogen phosphate
Formula:
NH₄H₂PO₄ MW: 115.03 g/mol Boiling Pt: 197 °C (dec.) Melting Pt: 190 °C Density: 1.803 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003396 CAS Number: 7722-76-1 EINECS: 231-764-5 |
Specification Test Results
Identity (IR) | complying |
Assay | Min. 98.0 % |
Water insoluble matter | Max. 0.005 % |
pH (5 %, 25 °C) | 3.8 - 4.4 |
Calcium (Ca) | Max. 0.001 % |
Iron (Fe) | Max. 0.001 % |
Potassium (K) | Max. 0.005 % |
Magnesium (Mg) | Max. 5 ppm |
Sodium (Na) | Max. 0.005 % |
Heavy metals (as Pb) | Max. 5 ppm |
Chloride (Cl) | Max. 5 ppm |
Nitrate (NO₃) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...