Ammonium chloride ≥99.5%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Ammonium muriate, Salmiac

31107-6X2.5KG 31107-6X500G 31107-6X1KG
BJ31107-1KGCS 752.3 USD
BJ31107-1KG BJ31107-2.5KG BJ31107-500G BJ31107-6X1KG
Ammonium chloride ≥99.5%, Puriss. p.a. Reag. Ph. Eur., ACS, ISO, Fluka™
Ammonium chloride
Formula: NH₄Cl
Melting Pt: 338 °C (dec.)
Density: 1.536 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011420
CAS Number: 12125-02-9
EINECS: 235-186-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e45c4909ae60875d5b9dca2f28f54f5c8028e64&vid=389479ce9f26d046837e12f5c47dd4bc7d47ec2b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay Min. 99.5 %
Assay (calculated to the dried substance) 99.5 - 100.5 %
Water insoluble matter Max. 0.005 %
Loss on drying (105°C) Max. 0.5 %
Residue on ignition (SO₄) Max. 0.01 %
pH (5 %, 25 °C) 4.5 - 5.5
Acidity or alkalinity complying
Arsenic (As) Max. 0.5 ppm
Calcium (Ca) Max. 5 ppm
Copper (Cu) Max. 2 ppm
Iron (Fe) Max. 2 ppm
Potassium (K) Max. 0.005 %
Magnesium (Mg) Max. 5 ppm
Vapor Pressure 1 mmHg ( 160.4 °C)
Sodium (Na) Max. 0.005 %
Nickel (Ni) Max. 0.0001 %
Lead (Pb) Max. 1 ppm
Zinc (Zn) Max. 0.0002 %
Heavy metals (ICP) ACS Max. 5 ppm
Nitrate (NO₃) Max. 0.0005 %
Phosphate (PO₄) Max. 2 ppm
Sulfate (SO₄) Max. 20 ppm
Iodide and bromide complying
Appearance of the solution complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR