Aluminium chloride hexahydrate 99%, Reagent Grade
Supplier: Honeywell Research Chemicals
|
Warning
|
237078-100G
237078-500G
BJ237078-100GEA
56
USD
BJ237078-100G
BJ237078-500G
Aluminium chloride hexahydrate 99%, Reagent Grade
Aluminium chloride hexahydrate
Formula:
AlCl₃·6H₂O Density: 2.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149134 CAS Number: 7784-13-6 |
Specification Test Results
Vapor Pressure | 1 mmHg ( 100 °C) |
Assay | 98.5 - 101.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...