Alizarin complexone ∼85% for the determination of fluorine, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: Alizarin-3-methyliminodiacetic acid, 2,2'-(((3,4-Dihydroxy-9,10-dioxo-9,10-dihydroanthracen-2-yl)methyl)azanediyl)diacetic acid, 3,4-dihydroxyanthraquinon-2-ylmethyliminodi(acetic acid), Alizarine-3-methylamine-N,N-diacetic acid

05590-1G 05590-5G
BJ05590-1GEA 280.35 USD
BJ05590-1G BJ05590-5G
Alizarin complexone ∼85% for the determination of fluorine, Fluka™
Alizarin complexone
Formula: C₁₉H₁₅NO₈
MW: 385.33 g/mol
Melting Pt: 180
Storage Temperature: Ambient
MDL Number: MFCD00001202
CAS Number: 3952-78-1
EINECS: 223-544-2

Order Now

Specification Test Results

Appearance complying
Assay 80.0 - 120.0 %
Appearance of the solution complying
Loss on drying (110°C) Max. 10 %
Absorption maximum 422 - 432 nm
Absorbtion (1%/1cm) Min. 75


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR