Acetic anhydride ≥99% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Acetyl oxideAcetyl acetate

33214-500ML 33214-6X1L 33214-4X2.5L
BJ33214-1LCS 855.35 USD
BJ33214-1L BJ33214-2.5L BJ33214-500ML
Acetic anhydride ≥99% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Fluka™
Acetic anhydride
Formula: (CH₃CO)₂O
MW: 102.09 g/mol
Boiling Pt: 140 °C
Melting Pt: -73.1 °C
Density: 1.083 g/cm³ (20 °C)
Flash Pt: 10.3 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00008705
CAS Number: 108-24-7
EINECS: 203-564-8
UN: 1715
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/22/53/2302253.jpg

Order Now

Specification Test Results

Assay (GC) Min. 99 %
Boiling range 136 - 138 °C
Explosion Limit 10.3 %
Density (D 20/4) 1.079 - 1.082
Non-volatile matter Max. 0.003 %
Aluminium (Al) Max. 0.5 ppm
Boron (B) Max. 0.02 ppm
Barium (Ba) Max. 0.1 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.05 ppm
Cobalt (Co) Max. 0.02 ppm
Chromium (Cr) Max. 0.05 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.1 ppm
Potassium (K) Max. 0.5 ppm
Lithium (Li) Max. 0.1 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.02 ppm
Molybdenum (Mo) Max. 0.1 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.1 ppm
Tin (Sn) Max. 0.1 ppm
Strontium (Sr) Max. 0.1 ppm
Zinc (Zn) Max. 0.2 ppm
Heavy metals (as Pb) Max. 0.0002 %
Chloride (Cl) Max. 0.0002 %
Phosphate (PO₄) Max. 0.001 %
Sulfate (SO₄) Max. 0.0005 %
KMnO₄ reducing matter (as O) Max. 0.01 %
Reaction against H₂SO₄ complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR