DSPE-PEG(2000) Maleimide ≥99%

Supplier: AVANTI POLAR LIPIDS INC MS
Danger

Synonyms: Poly(oxy-1,2-ethanediyl), α-[(9R)-6-hydroxy-6-oxido-1,12-dioxo-9-[(1-oxooctadecyl)oxy]-5,7,11-trioxa-2-aza-6-phosphanonacos-1-yl]-ω-[2-[[3-(2,5-dihydro-2,5-dioxo-1H-pyrrol-1-yl)-1-oxopropyl]amino]ethoxy]-, ammonium salt (1:1)1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[maleimide(polyethylene glycol)-2000] (ammonium salt)DSPE-PEG2000-Mal ammonium

880126P25MG 880126P500MG 880126P100MG 880126P50MG 880126P10MG
100127-908EA 1221.54 USD
100127-908 100120-480 100121-480 100125-680 100124-602
DSPE-PEG(2000) Maleimide ≥99%
DSPE-PEG(2000) Maleimide
MW: 2941.605 g/mol
Storage Temperature: Freezer
CAS Number: 474922-22-0
UN: 1888
ADR: 6.1,III

Order Now

Specification Test Results

Purity >99%
Light Sensitive No
Hygroscopic Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR