The Apprentice Doctor® Budget Phlebotomy Training Arm
Supplier: THE APPRENTICE CORPORATION
AD604R-LIGHT AD604R-DARK AD603-BFN10
470313-860CS 904.4 USD
470313-860 470313-862 470355-598
The Apprentice Doctor® Budget Phlebotomy Training Arm
Educational Materials Biology Educational Materials Human Biology Learning Activities
Priced so everyone can have their own.
  • Learn or refresh Venipuncture, Phlebotomy, and IV skills
  • Gain confidence as a medical professional
  • Demonstrate what to do to others
Offering a high level of realism, this simulation arm has the anatomically accurate venous anatomy for both dorsal (back of the hand) and ventral (cubital fossa) surfaces. The veins are palpable and partially visible. They provide a give-feel effect when entering the vein, so users feel like they are treating a live patient. The realistic touch and feel of the phlebotomy training armMore Product Information