SPE Columns for THC, Honeywell Burdick & Jackson
Supplier: Honeywell Burdick & Jackson
9910 9918 9912 9916 9914
BJ9916PK 286.56 USD
BJ9916 BJ9918 BJ9912 BJ9914 BJ9910
SPE Columns for THC, Honeywell Burdick & Jackson
Chromatography Columns
These SPE columns are made from medical-grade polypropylene with PE frits.
  • Every stationary phase batch tested to ensure lot-to-lot reproducibility
  • Reliable quality