Soil Extract Agar, HiMedia Laboratories
Supplier: HiMedia
M455-500G M455-2.5KG M455-5KG
95021-852EA 232.07 USD
95021-852 89225-574 89225-576
Soil Extract Agar, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Soil extract agar is used for isolation of soil microorganisms.
  • Cream to yellow homogeneous free flowing powder
  • Firm gelling, comparable with 1.5% agar gel
  • Cultural characteristics can be observed after an incubation at 30 °C for up to 4 days