Magnesium sulfate, anhydrous ≥99.5%, Reagent Grade

Supplier: Honeywell Research Chemicals

Synonyms: MgSO4Magnesium sulfate anhydrousMagnesium sulphate anhydrous

M7506-500G
BJM7506-500GEA 151.94 USD
BJM7506-500G
Magnesium sulfate, anhydrous ≥99.5%, Reagent Grade
Magnesium sulphate
Formula: MgSO₄
MW: 120.37 g/mol
Melting Pt: 1.130 °C (dec.)
Density: 2.7 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011110
CAS Number: 7487-88-9
EINECS: 231-298-2
https://us.vwr.com/stibo/web/std.lang.all/06/17/16640617.jpg

Order Now

Specification Test Results

Assay (calculated to the dried substance) Min. 99.5 %
Loss on drying (105°C) Max. 2 %
Colour of the solution complying
Clearness of the solution complying

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR