1,2-Dichloroethane ≥99.0% ACS
Supplier: AQUA SOLUTIONS MS
|
Danger
|
Synonyms:
Ethylene chloride, Ethylene dichloride, Dichloroethane
D4490-20L
D4490-4L
D4490-1L
D4490-500ML
101225-076EA
726.86
USD
101225-076
101224-992
101224-990
101226-114
1,2-Dichloroethane ≥99.0% ACS
1,2-Dichloroethane
Formula:
ClH₂CCH₂Cl MW: 98.96 g/mol |
MDL Number:
MFCD00000963 CAS Number: 107-06-2 EINECS: 203-458-1 UN: 1184 ADR: 3,6.1,II |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...