Tartrazine 1.5% in aqueous solution for bacteriology (Brown-Hopps method)
Supplier: Electron Microscopy Sciences
|
Warning
|
Synonyms:
FD and C Yellow No. 5
, Sodium 5-oxo-1-(4-sulphonatophenyl)-4-((4-sulphonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylate
, Acid Yellow 23
, Sodium 5-oxo-1-(4-sulfonatophenyl)-4-((4-sulfonatophenyl)diazenyl)-4,5-dihydro-1H-pyrazole-3-carboxylate
, Filter Yellow
26106-06
100504-506EA
41.39
USD
100504-506
Tartrazine 1.5% in aqueous solution for bacteriology (Brown-Hopps method)
Tartrazine
Brown-Hopps Method reagent for Gram Positive and Gram Negative Bacteria.
Stain results:
Gram Negative Bacteria: Red
Gram Positive Bacteria: Blue
Background: Yellow
Formula:
C₁₆H₁₂N₄O₉S₂ MW: 534.36 g/mol |
MDL Number:
MFCD00148908 CAS Number: 1934-21-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...