Basic fuchsin 0.5% in aqueous solution staining solution, for bacteriology (Brown-Hopps method)
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Basic-fuchsin
, Diamondfuchsin
, Basic Red 9
, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride
, Basic fuchsin
, Fuchsine base monohydrochloride
, Rosaniline hydrochloride
, Pararosaniline
, Fuchsin basic
, Basic fuchsin hydrochloride
, CI 42510
, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride
, Magenta O
Brown-Hopps Method reagent for Gram Positive and Gram Negative Bacteria.
Stain results:
Gram Negative Bacteria: Red
Gram Positive Bacteria: Blue
Background: Yellow
Formula:
C₂₀H₂₀ClN₃ MW: 337.85 g/mol |
MDL Number:
MFCD00012569 CAS Number: 632-99-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...