Tetrabutylammonium hydroxide 0.98-1.02 M in water for HPLC, for ion pair chromatography, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger


Management of Change (MOC) category = N

BAKR9580-03 BAKR9580-03
JT9580-3EA 11075.48 USD
CAJT9580-3 JT9580-3
Tetrabutylammonium hydroxide 0.98-1.02 M in water for HPLC, for ion pair chromatography, J.T.Baker®
Tetrabutylammonium hydroxide
Formula: C₁₆H₃₇NO
MW: 259.48 g/mol
Density: 0.83 g/cm³
MDL Number: MFCD00009425
CAS Number: 2052-49-5
UN: 3267
ADR: 8,III

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay((C₄H₉)₄NOH), Molarity 0.98 - 1.02
Ultraviolet Absorbance (0.1M Solution in Water) - 240 nm ≤0.15
Ultraviolet Absorbance (0.1M Solution in Water) - 245 nm ≤0.10
Ultraviolet Absorbance (0.1M Solution in Water) - 254 nm ≤0.07
Ultraviolet Absorbance (0.1M Solution in Water) - 280 nm ≤0.04
Ultraviolet Absorbance (0.1M Solution in Water) - 300 nm ≤0.04

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR