Polyflow®-G Polypropylene Nominal-Rated Depth Cartridges, Parker Hannifin
Supplier: PARKER HANNIFIN TE
PG-10010-010-14 PG-10010-050-14 PG-10010-100-14 PG-10110-050-11 CO-10020 PG-10310-002-1 PG-10310-010-2 PG-10320-005-1 PG-10320-010-1 PG-10330-005-1 PG-10620-002-0 PG-10620-010-0 PG-10710-002-2 PG-10H20-005-1 PG-10010-100-12
76239-136CS 1080.1 USD
76239-136 76239-138 76239-140 76239-142 76239-144 76239-146 76239-148 76239-150 76239-152 76239-154 76239-156 76239-158 76239-160 76239-162 76239-164 76240-414
Polyflow®-G Polypropylene Nominal-Rated Depth Cartridges, Parker Hannifin
Filters Filter Cartridges
These are for economical prefiltration. Polyflow®-G depth media has been developed for a wide variety of general process applications from fluid clarification to general prefiltration. Its high dirt-loading, random-fiber depth media provides consistent particle retention.
  • Thermally bonded from 100% virgin polypropylene to ensure clean filtrates and excellent chemical and thermal compatibility
  • Overall reduction of filtration costs when compared to spunbonded, stringwound, and nominally-rated pleated prefilter cartridges
  • Longer filtration life reduces downtime due to fewer change-outs
High flow rate and long service life reduce processing time. Broad chemical compatibility allows use in most applications. Thermally bonded construction minimizes extractables for cleaner filtrate.
Applied for liquid clarification, general water filtration, beverage/wine clarifi cation, RO/DI prefiltration.
Effective filtration area: 0.33 m² (3.6 ft²) per 250 mm (10") cartridge.
Cartridge extractables: NVR <35 mg per 250 mm (10") cartridge.
Maximum differential pressure/temperature of forward: 80 psid (5.5 bar) at 75 °F (24 °C) and reverse: 40 psid (2.8 bar) at 75 °F (24 °C), 15 psid (1.0 bar) at 140 °F (60 °C).
Certifications: Meets USP specifi cations for Class VI-121 °C plastics criteria.