Crystal Polishing Kit, PerkinElmer
Supplier: PerkinElmer
51001188
10139-174EA 1163.46 USD
10139-174
Crystal Polishing Kit, PerkinElmer
Polishers and Tumblers
Fogged windows are quickly restored with the infrared crystal polishing kit.
  • Special self-adhesive selvyts keeps the selvyt flat and makes replacement quick and simple
The kit contains everything necessary to repolish sodium chloride, potassium bromide, or cesium iodide windows.
Delivery information: Kit includes a smoothing lap, polishing rouge, a sponge, an optical flat, a Selvyt cloth, and a booklet describing the repolishing technique. Special self-adhesive selvyts that stick to the polishing lap are also supplied.