Environmental Express Digestion Cups, Cole Parmer
Supplier: Cole Parmer
53203-13 53204-17
MSPP-EW5320417PK 138.46 USD
MSPP-EW5320417 MSPP-EW5320313
Environmental Express Digestion Cups, Cole Parmer
Digestion Systems Mineralization Workstations
The cups that act as a digestion vessel or storage container and fit perfectly into the HotBlock and HotBlock Pro systems.
  • Low metals background and EPA approved for digestions
  • Redesigned for optimum performance
Wobble free deeper bottom dimple provides added stability
Delivery information: It includes linerless caps, and certificate of volume accuracy and metals background analysis report for 68 elements to both ppb and ppt (parts per trillion) levels.