Nitric acid fuming ≥90%, ChemPUR® ACS
Supplier: Puritan Products, Inc.
|
Danger
|
2713871
2713964
76266-906EA
931.73
USD
76266-906
76266-908
76267-082
76267-084
76267-086
76267-088
76267-090
76267-092
Nitric acid fuming ≥90%, ChemPUR® ACS
Nitric acid
Formula:
HNO₃ |
MDL Number:
MFCD00011349 CAS Number: 7697-37-2 UN: 2031 ADR: 8,I |
Specification Test Results
Dissolved Oxides | Max.0.1% |
Assay (w/w) | Min. 90% |
Sulfate | Max. 5.0 ppm |
Dilution Test | Passes Test |
Residue after Ignition | Max. 20 ppm |
Chloride | Max. 0.7 ppm |
Arsenic | Max. 0.3 ppm |
Heavy Metals | Max. 5.0 ppm |
Iron | Max. 2 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...