D-(+)-Glucose, anhydrous ACS

Supplier: AQUA SOLUTIONS MS

D2302-2.5KG D2302-12KG D2302-500G
101223-960EA 516.55 USD
101223-960 101225-140 101226-366
D-(+)-Glucose, anhydrous ACS
D(+)-Glucose

Dextrose (Glucose) Anhydrous Reagent ACS Grade


  • Appearance White powder
  • Harmonization Code 1702.60.4000

Formula: C₆H₁₂O₆
MDL Number: MFCD00148912
CAS Number: 50-99-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47ec28c7fd4803097405f8eaef3171b3a40d40c5&vid=62627c96e2ae9f2a0cae732d0ddf0a100247298e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Insoluble Matter 0.005%
Loss on Drying 0.2%
Residue after Ignition 0.02%
Titrable Acid 0.002 meq/g
Chloride (Cl) 0.01%
Sulfate and Sulfite (as SO4) 0.005%
Heavy Metals (as Pb) 5 ppm
Starch Passes test
Iron (Fe) 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR