Congo red 0,2% alcoholic solution
Supplier: Electron Microscopy Sciences
|
Danger
|
Synonyms:
Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulphonate), Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulphonate), Disodium 3,3'-[[1,1'-biphenyl]-4,4'-diylbis(azo)]bis(4-aminonaphthalene-1-sulfonate), CI 22120, Sodium 3,3'-([1,1'-biphenyl]-4,4'-diylbis(diazene-2,1-diyl))bis(4-aminonaphthalene-1-sulfonate), Direct Red 28
26090-06
26090-05
100504-486EA
106.2
USD
100504-486
101411-004
Congo red 0,2% alcoholic solution
Congo Red
Formula:
C₃₂H₂₂N₆Na₂O₆S₂ Boiling Pt: 78 °C Flash Pt: 13 °C |
MDL Number:
MFCD00004028 CAS Number: 573-58-0 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...