Leakproof HDPE Bottles, Natural, C&G Containers
Supplier: C&G Containers
LPV008249002 LPV008200052
76313-900CS 127.99 USD
76313-900 76313-978
Leakproof HDPE Bottles, Natural, C&G Containers
Bottles Plastic Bottles
These leakproof HDPE bottles are used for aqueous sample collection and for testing metals and a variety of inorganic parameters including alkalinity, anions, acidity, cyanide, sulfate, and hardness.
Wash protocols for these bottles include, A1 (Nitric acid wash), B1 (Deionized water wash), C1 (Hydrochloric acid wash), and L3 (no wash). Some bottles are chemically preserved.
Pre-cleaned and Certified products contain the Certificate of Analysis and Cleaning Protocol Form in the product case.
A lot certificate of analysis is generated with the analytical results obtained includingMore Product Information