Water USP for irrigation, sterile
Supplier: B BRAUN MEDICAL
Synonyms:
Dihydrogen oxide
R8005
76313-486CS
244.25
USD
76313-486
Water USP for irrigation, sterile
Water
Sterile Water of Irrigation USP is a sterile, hypotonic, nonpyrogenic irrigating fluid or pharmaceutic aid (solvent) entirely composed of Sterile Water for Injection USP.
- Innovative, convenient port system
- Tamper-evident injection site
- Highly transparent
It is prepared by distillation and contains no antimicrobial or bacteriostatic agents or added buffers.
Not made with natural rubber latex, DEHP or PVC.
NDC number: 00264-7386-50
Formula:
H₂O MW: 18.02 g/mol |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...