Water USP for injection (WFI), sterile

Supplier: B BRAUN MEDICAL

Synonyms: Dihydrogen oxide

L8500
32500-211CS 74.67 USD
32500-211
Water USP for injection (WFI), sterile
Water

Sterile Water for Injection USP is a clear, colorless, odorless liquid. It is sterile, hypotonic, nonpyrogenic, and contains no bacteriostatic or antimicrobial agents. It is a diluent or solvent suitable for intravascular injection after first having been made approximately isotonic by the addition of suitable solute.


  • NDC number: 0264-7850-00


Sterile Water for Injection USP is also used as a diluent or solvent for other parenteral drugs. As such, it contributes to hydration when provided in parenteral drug and fluid therapy, after the introduction of suitable additives and/or mixture with suitable solutes to approximate isotonicity.


Not manufactured with latex, PVC or DEHP.

Formula: H₂O
MW: 18.02 g/mol
MDL Number: MFCD00011332
CAS Number: 7732-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR