Sodium chloride 0,45% USP for injection

Synonyms: Saline, NaCl, Sea salt, Halite

L8020 L8021
76313-482CS 100.69 USD
76313-482 76313-484
Sodium chloride 0,45% USP for injection
Sodium chloride

0.45% Sodium Chloride Injection USP is primarily a hydrating solution and may be used to assess the status of the kidneys, since more water is provided than is required for excretion of salt.


It may also be used in the treatment of hyperosmolar diabetes where the use of dextrose is inadvisable and there is a need for large amounts of fluid without an excess of sodium ions.


Not manufactured with latex, PVC or DEHP.


Specific Gravity: 1.003


NDC number: 00264-7802-00

Formula: NaCl
MW: 58.44 g/mol
Boiling Pt: 1413
Melting Pt: 801
MDL Number: MFCD00003477
CAS Number: 7647-14-5
Merck Index: 14,08599

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR