Tris(dibenzylideneacetone)dipalladium(0) ≥97.0% (carbon basis)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Pd2(dba)3, Pd2dba3, Tris(dibenzylideneacetonyl)bis-palladium, Bis[tris(dibenzylideneacetone)palladium(0)]

04904-5G 04904-1G 04904-25G 04904-1G
102566-068EA 380 USD
102566-068 102619-288 102619-290 CH04904-1G
Tris(dibenzylideneacetone)dipalladium(0) ≥97.0% (carbon basis)
Tris(dibenzylideneacetone)dipalladium(0)

Catalyst for stille reactions of support bound stannane.

Formula: C₅₁H₄₂O₃Pd₂
MW: 915.72 g/mol
Melting Pt: 152...155 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00013310
CAS Number: 51364-51-3

Order Now

Specification Test Results

Appearance Purple Brown Microcrystalline Powder
Elemental Analysis (Found), C 66.89 ±1.7%
Elemental Analysis (Found), H 4.62 ±0.6%
Infrared Spectrum Conforms to Structure
Assay 20.00 to 24.2% (Palladium Content)
Purity ≥97.0% (Based on Carbon Content)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR