tert-Butyl-5-amino-1H-indazole-1-carboxylate ≥98% (by HPLC)

Supplier: CHEM IMPEX INTL INC MS

Synonyms: tert-Butyl 5-amino-1H-indazole-1-carboxylateN-(1)-Boc-5-aminoindazole5-Amino-1H-indazole-1-carboxylic acid, 1,1 dimethyl ethyl ester5-Amino-indazole-1-carboxylic acid tert-butyl ester1-Boc-5-amino-indazole

17593-1G 17593-5G
102642-188EA 53.57 USD
102642-188 102622-676
tert-Butyl-5-amino-1H-indazole-1-carboxylate ≥98% (by HPLC)
tert-Butyl-5-amino-1H-indazole-1-carboxylate
Formula: C₁₂H₁₅N₃O₂
MW: 233.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114656
CAS Number: 129488-10-4

Order Now

Specification Test Results

Appearance Grey to Yellow or White Solid
¹H NMR Conforms to Structure
Purity by HPLC ≥98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR