Klinger Screw Dislocation Crystal Model
Supplier: Klinger
Ward's®
KS8109
470311-158EA 750.05 USD
470311-158
Klinger Screw Dislocation Crystal Model
Models Crystal Models
This model is hand made in the USA by Klinger Educational Products.
This is a permanent structure. We only use grade A materials. The 1" balls are made of hard Maplewood that includes an enameled painted finish. Polished steel rods are used to connect the wooden balls together.