MicroFil™ Pipette Filler, World Precision Instruments
Supplier: World Precision Instruments
MF28G67-5
MSPP-MF28G675PK 116.31 USD
MSPP-MF28G675
MicroFil™ Pipette Filler, World Precision Instruments
Pipettes Pipette Controllers
This long and fine tip allows to start the filling very close to the pipette tip, eliminating both air bubble formation and clogging due to the washing down of dust particles.
  • Fills micropipettes easily and reliably
  • Stored for days with the filling solution inside without clogging
  • Tip elasticity is sturdy and very flexible though not unbreakable
  • More flexible than stainless steel needles
  • Luer fitting allows easy coupling to syringes and syringe filters
The transparent MicroFil™ needle is constructed from a combination of plastic and fused silica - no metal components are used. The Luer fitting is HDPP and is held in place with a medical grade UV adhesive.