Sodium hydroxide 10 mol/L, EMPROVE® EXPERT, SAFC®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

Filtered through a 0.2 µm filter.

4.80648.9025 4.80648.9950 4.80648.2500 4.80648.4000 4.80648.9208 4.80648.9958
EM4.80648.9025EA 1394.45 USD
EM4.80648.9025 EM4.80648.9950 EM4.80648.2500 EM4.80648.4000 EM4.80648.9208 EM4.80648.9958
Sodium hydroxide 10 mol/L, EMPROVE® EXPERT, SAFC®
Sodium hydroxide

Store above +15 °C.


This product has been manufactured from Sodium hydroxide pellets Ph. Eur., BP, JP, NF and water for injection (wfi) acc. USP which conforms with highly purified water (hpw) acc. Ph. Eur.


Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390
Melting Pt: 323
Density: 1.33 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8 II
Merck Index: 14,08627

Order Now

Specification Test Results

Assay (acidimetric,NaOH) 9.9 - 10.1 mol/l
Assay (acidimetric) 29.9 - 30.3 %
Identity passes test
Carbonate (as Na₂CO₃) ≤ 1.0 %
Chloride (Cl) ≤ 20 ppm
Sulfate (SO₄) ≤ 30 ppm
Heavy metals (as Pb) ≤ 5.0 ppm
Al (Aluminium) ≤ 10 ppm
Fe (Iron) ≤ 3 ppm
Ni (Nickel) * ≤ 2.00 ppm
Residual solvents (ICH Q3C) ...excluded by production process
Endotoxins ≤ 2.0 I.U./ml


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR