Biuret ≥98%

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Allophanic acid amideCarbamylurea

22844-25G 22844-100G 22844-250G
102581-262EA 36.54 USD
102581-262 102598-880 102598-878
Biuret ≥98%
Biuret
Formula: C₂H₅N₃O₂
MW: 103.08 g/mol
Melting Pt: 182...197 °C
Storage Temperature: Ambient
MDL Number: MFCD00007946
CAS Number: 108-19-0
https://us.vwr.com/stibo/web/std.lang.all/33/26/2353326.jpg

Order Now

Specification Test Results

Appearance White Crystalline Powder
Infrared Spectrum Conforms to Structure
Loss on Drying ≤2.5%
Assay ≥98%
Purity by TLC ≥99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR