Calcium chloride, anhydrous ≥93.0%, granular ≤7.0 mm, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

C1016-100G C1016-2.5KG C1016-500G
BJC1016-100GEA 65 USD
BJC1016-100G BJC1016-2.5KG BJC1016-500G
Calcium chloride, anhydrous ≥93.0%, granular ≤7.0 mm, Fluka™
Calcium chloride
Formula: CaCl₂
MW: 110.98 g/mol
Boiling Pt: 1600
Melting Pt: 772
Density: 2.150 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8
REACH: 01-2119494219-28
Merck Index: 14,01659

Order Now

Specification Test Results

Form granular
Assay Min. 93.0 %
Colour of the solution complying
Clearness of the solution complying
Granulation complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR