GFAAS Mixed Standard, Matrix per Volume 5% HNO₃, PerkinElmer
Supplier: Perkin Elmer
N9300244
97011-978EA 516.03 USD
97011-978
GFAAS Mixed Standard, Matrix per Volume 5% HNO₃, PerkinElmer
Standards Cationic Standards Multi-Element Standards
The mixed standard, matrix per volume 5% HNO₃, concentration of elements are 5 mg/l Beryllium and Cadmium, 10 mg/l Silver, 20 mg/l Chromium, Iron and Magnesium, 50 mg/l Barium, Cobalt, Copper and Nickel, 100 mg/l Aluminium, Arsenic, Lead, Antimony, Selenium and Thallium.