Sulfuric acid 0.02 N
Supplier: AQUA SOLUTIONS MS
|
Danger
|
9020-1L
9020-20L
9020-4LC
9020-10L
9020-500ML
9020-4L
101226-898EA
22.94
USD
101226-898
101225-424
101223-530
101223-778
101226-826
101223-114
Sulfuric acid 0.02 N
Sulfuric acid
Formula:
H₂SO₄ MW: 98.08 g/mol |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 |
Specification Test Results
Prepared to Formulation on File | X |
N.I.S.T. Traceable | X |
Actual Normality | 0.0200 ± 0.0005 Normal @ 20°C |
Appearance and Color | Clear water white liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...