Trident 286, 18" Natural Rubber Gloves, Mapa Professional
Supplier: MAPA
286310 286311 286319
75794-970CS 801.19 USD
75794-970 75795-032 75795-034
Trident 286, 18" Natural Rubber Gloves, Mapa Professional
Gloves
These natural rubber gloves has mechanical performance for long-lasting chemical protection.
  • Forearm protection
  • Chlorinated interior
  • Reinforced grip for better handling
Thickness of the material provides good impact protection, as well as a longer service life.
Certifications: CE Cat. III marked. EN 374: ABKL. EN 388: 2131