Ultrane 500 Grip and Proof Gloves, Mapa Professional
Supplier: MAPA
500418 500419 500410 500411 500417
10811-816CS 571.21 USD
10811-816 10811-874 10811-876 10811-878 10811-880 10811-906 10811-908 10811-910 10811-912 10811-914 10811-916 10811-918 10811-920 10811-922 10811-924
Ultrane 500 Grip and Proof Gloves, Mapa Professional
Gloves
With high fingertip sensitivity and assured grip, this glove was made for precision work in wet and oily environments.
  • Lightweight
  • Oilproof; repels oil penetration
  • Excellent abrasion resistance
  • Seamless liner
  • Washable until 5 cleaning cycles
Nitrile coating protects skin up to knuckles and ensures an excellent grip in oily conditions. Designed to reduce muscle strain and boost productivity.
Certifications: CE Cat. II marked. ISO 13997:3.4N (327g). ASNI CUT A1.