1-Heptanesulfonic acid sodium salt, anhydrous ≥99% (by titrimetric analysis) for HPLC

Supplier: CHEM IMPEX INTL INC MS

Synonyms: Sodium 1-heptanesulfonateSodium 1-heptanesulphonateSodium heptane-1-sulfonateSodium heptane-1-sulphonate

22841-250G 22841-5G 22841-100G 22841-1KG 22841-25G
102581-260EA 52.48 USD
102581-260 102624-322 102598-876 102598-874 102624-324
1-Heptanesulfonic acid sodium salt, anhydrous ≥99% (by titrimetric analysis) for HPLC
1-Heptanesulphonic acid sodium salt

It is HPLC reagent used for ion-association in analyses of peptides and proteins.

Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202.25 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00007543
CAS Number: 22767-50-6
https://us.vwr.com/stibo/web/std.lang.all/07/91/2300791.jpg

Order Now

Specification Test Results

Appearance White Crystalline Powder
pH 5.5 - 7.5 (10% in water)
Infrared Spectrum Conforms to Structure
UV Ratios, Transmittance (0.005 mol. aqueous solution) at 200 nm ≥70%
UV Ratios, Transmittance (0.005 mol. aqueous solution) at 220 nm ≥90%
UV Ratios, Transmittance (0.005 mol. aqueous solution) at 250 nm ≥98%
Loss on Drying ≤2% at 120 °C
Assay by Titration ≥99% (anhydrous basis)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR