Edge® 48-703 Industrial Work Gloves with Leather Palm, Ansell
Supplier: Ansell Healthcare
48703070 48703080 48703090 48703100 48703110
76196-278CS 1788.17 USD
76196-278 76196-280 76196-282 76196-292 76196-294
Edge® 48-703 Industrial Work Gloves with Leather Palm, Ansell
Gloves
Advanced cut and abrasion protection combined with premium leather for a superior grip.
  • Good dexterity and flexibility
  • Medium cut protection and high levels of abrasion protection
  • Good grip in dry and slightly oily environments
  • Good sensitivity, fit, and cushioning
  • Polyethylene/glass fiber liner
Made with a seamless liner as well as a knitted wrist to offer close fit in cut resistant applications. Combines a good level of cut, abrasion, and thermal resistance in one glove. The leather palm re-enforcement provides extra protection for the worker in cut and hot parts handling.
Certifications: CE 0493 marked. EN 388:2003 - 4543 mechanical hazards. EN 388:2016 - 4X43D mechanical hazards. EN 407 - X1XXXX heat protection. REACH Complaint.