ActivArmr® 97-681 Cold and Liquid Protection Gloves, Ansell
Supplier: Ansell Healthcare
97681080 97681090 97681100 97681110
76196-270CS 600.02 USD
76196-270 76196-272 76196-274 76196-276
ActivArmr® 97-681 Cold and Liquid Protection Gloves, Ansell
Gloves
Industry leading combination of warmth and dexterity in a liquid-proof glove. Thin enough to handle small objects but protective enough for harsh environments, this glove helps wearers outsmart the elements.
  • PVC foam coating stays flexible down to –25 °C/–15 °F
  • Helps to fight frostbite
  • Water-proof Ripel™ technology
  • Food safe
Multi-level construction takes into account all aspects of hand protection when working conditions are cold and wet. The PVC coating provides grip and flexibility, even in the coldest temperatures. Ripel™ technology keeps liquids out, helping hands stay dry and warm longer.
The interior is made of a unique two layer design; flexible nylon to protect against liquid and mechanical risks, and soft, insulative acrylic to provide comfort and warmth. ActivArmr® is unlike anything in the market by giving the wearer a unique combination of warmth, dexterity, and flexibility that lets users feel everything but the cold.
Certifications: ANSI level 3 abrasion resistance. ANSI level A2 cut protection. EN 388:2016 - 2231B mechanical hazards. EN 511 - 221 cold protection. CE marked. Food safe. REACH Compliant.