PBI/Kevlar® High Heat Glove with Wing Thumb, Steel Grip
Supplier: Steel Grip
GGPB302449-14PA
80701-466PR 335.8 USD
80701-466
PBI/Kevlar® High Heat Glove with Wing Thumb, Steel Grip
Gloves
Perfect for high heat applications, this glove is constructed with a PBI/Kevlar® blend, a thermonol cuff, Kevlar® sewn thread, and an additional Nomex® aramid palm patch for unbeatable protection.
  • 30 oz. PBI/KEVLAR® blend
  • 22 oz. thermonol cuff
  • 14 oz. wool lining
Glove color: Tan