Sodium thiosulfate 0.025 N

Supplier: AQUA SOLUTIONS MS

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

8800-4LC 8800-4L 8800-500ML 8800-20L 8800-10L 8800-1L
101223-546EA 60.95 USD
101223-546 101223-888 101223-386 101225-414 101225-776 101227-060
Sodium thiosulfate 0.025 N
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
MDL Number: MFCD00003499
CAS Number: 7772-98-7
Merck Index: 12,08844

Order Now

Specification Test Results

Actual Normality 0.0245 - 0.0255


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR