Sulfuric acid solution 10 N

Supplier: AQUA SOLUTIONS MS
Danger

9106-1L 9106-20L 9106-500ML 9106-4L 9106-4LC
101224-696EA 349.39 USD
101224-696 101223-700 101223-698 101223-206 101223-332
Sulfuric acid solution 10 N
Sulfuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
MDL Number: MFCD00064589
CAS Number: 7664-93-9
UN: 2796
ADR: 8,II

Order Now

Specification Test Results

Prepared to Formulation on File Yes
Appearance and color Clear water white liquid
Normality 9.950 - 10.050
N.I.S.T. Traceable to SRM 84 Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR