Empty Micro SpinColumns, Harvard Apparatus
Supplier: Harvard Apparatus
744421 744431 744401 744400
101166-922PK 206.15 USD
101166-922 101166-930 101166-920 101166-926
Empty Micro SpinColumns, Harvard Apparatus
Filters Centrifugal Filters
Empty QuikPrep® SpinColumns™ provide the flexibility to fill columns with own chromatographic material.
  • Can be prefilled with custom material