Chicken Embryo Extract, MP Biomedical

Supplier: MP Biomedicals
092850145
IC092850145EA 592.21 USD
IC092850145
Chicken Embryo Extract, MP Biomedical
Cell Culture Media
Chicken embryo extract is widely used as cell culture supplement for avian neural crest cells, muscle cells, dorsal root ganglion neurons etc. The product is supplemented with 100 IU/ml penicillin, 100 µg/ml streptomycin and 2.5 µg/ml amphotericin, and sterile filtered to 0.2 µm.

  • Sterile
  • Filtered to 0.2 µm
  • Soluble in water
  • Typically used at a concentration of 0.1% to 2% in cell culture

Chicken embryo extract is prepared from 9 to 12 day old eggs collected from registered flocks and found to be free from clinical signs of notifiable diseases.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR