MicroMaxNS Disposable Protective Coveralls with Elastic Wirsts and Ankles, Zipper Closure, Lakeland Industries
Supplier: Lakeland
CTL417-2X CTL417-XL CTL417-3X CTL417-4X
76246-316CS 339.29 USD
76246-316 76309-064 76309-066 76309-068 76309-070 76309-072 76310-152 76310-154
MicroMaxNS Disposable Protective Coveralls with Elastic Wirsts and Ankles, Zipper Closure, Lakeland Industries
Disposable Garments
These disposable protective coveralls are made from lightweight microporous fabric.
  • Microporous film laminated to polypropylene substrate
  • Serged seams
  • Blood-borne pathogen/viral barrier tested
MicroMax NS coverall offers the protection against dirt, dust, grease, grime and light chemical splash.
Elastic wrists and ankles for better fit and increased protection.