MicroMax® NS Disposable Protective Coveralls with Elastic Wrists and Attached Hood and Boots, Zipper Closure, Lakeland Industries
Supplier: Lakeland
CTL414-4X CTL414-5X
75856-882CS 575.97 USD
75856-882 76309-034 76309-036 76309-038 76309-040 76309-042 76309-044 76309-046
MicroMax® NS Disposable Protective Coveralls with Elastic Wrists and Attached Hood and Boots, Zipper Closure, Lakeland Industries
Disposable Garments
Protection from dirt, dust, grease, grime, and light chemical splash.
  • Economical
  • Light weight
  • Features high MVTR (Moisture Vapor Transmission Rate)
  • Material: 55 g SBPP with laminated microporous film
MicroMax® NS is a line of general-purpose protective clothing that can be used in work environments where hazardous or nonhazardous contaminants may be present. Very economical, lightweight, and flexible, these garments provide a high Moisture Vapor Transmission Rate (MVTR) for added wearer comfort. MicroMax® NS brand fabric is comprised of a laminated microporous film and is strong, wet or dry, perfect for work environments where dirt, grime, splashes, and spills are present.
Certifications: ASTM F1670. ASTM F1671.