Egg PC ≥99%

Supplier: AVANTI POLAR LIPIDS INC MS

Synonyms: L-α-phosphatidylcholine (Egg, Chicken), Phosphatidylcholine from egg yolk

840051C200MG 840051C25MG 840051C500MG 840051C1G
100123-724EA 295.38 USD
100123-724 100127-780 100120-836 100128-404
Egg PC ≥99%
Egg PC

L-α-phosphatidylcholine (Egg, Chicken), chloroform


  • Concentration 10 mg/mL (25mg)
  • Concentration 25 mg/mL (200mg, 500mg, 1g)

Storage Temperature: Freezer
CAS Number: 97281-44-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR