Egg PC 25 mg/mL in chloroform ≥95%

Supplier: 0000018221

Synonyms: L-α-phosphatidylcholine (Egg, Chicken)Phosphatidylcholine from egg yolk

131601C1G
100120-632EA 585.32 USD
100120-632
Egg PC 25 mg/mL in chloroform ≥95%
Egg PC

L-α-phosphatidylcholine (95%) (Egg, Chicken), chloroform


Egg PC is a further purification of Total Egg Extract. This Natural Lipid is a mixture, and the structure shown above is only representative of one possible structure present in the product. Refer to chart for average fatty acid distribution.

Storage Temperature: Freezer
CAS Number: 97281-44-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR